Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EMX1 Polyclonal Antibody
Chicken Polyclonal Antibody
Supplier: Invitrogen™ PA572742
Description
EMX1 Polyclonal Antibody for Western Blot, ELISA
EMX1 is a transcription factor, which in cooperation with EMX2. It acts to generate the boundary between the roof and archipallium in the developing brain. It may function in combinations with OTX1/2 to specify cell fates in the developing central nervous system.
Specifications
EMX1 | |
Polyclonal | |
Unconjugated | |
Emx1 | |
empty spiracles homeobox 1; empty spiracles homolog 1; Empty spiracles-like protein 1; EMX1; Emx-1; Homeobox protein EMX1; RGD1564002 | |
Chicken | |
Antigen affinity chromatography | |
RUO | |
13796, 2016, 500235 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q04741, Q04742 | |
Emx1 | |
EMX1 antibody for a 16 amino acid synthetic peptide near the amino terminus of human EMX1. The immunogen is within amino acids 50 - 100 of EMX1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgY |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction