Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EMX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595747
Description
Immunogen sequence: AAAASEEPLR PTALNYPHPS AAEAAFVSGF PAAAAAGAGR SLYGGPELVF PEAMNHPALT VHPAHQLGAS PLQPPHSFFG AQHRDPLHFY PWVLRNRFFG HRFQASDVPQ DGLLLHGPFA R; Positive Samples: Rat liver; Cellular Location: Cytoplasm, Nucleus.
EMX1 is a transcription factor, which in cooperation with EMX2. It acts to generate the boundary between the roof and archipallium in the developing brain. It may function in combinations with OTX1/2 to specify cell fates in the developing central nervous system.
Specifications
EMX1 | |
Polyclonal | |
Unconjugated | |
Emx1 | |
Emx1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 70-190 of human EMX1 (NP_0040882). | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
2.03 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
empty spiracles homeobox 1; empty spiracles homolog 1; Empty spiracles-like protein 1; EMX1; Emx-1; Homeobox protein EMX1; RGD1564002 | |
Rabbit | |
Affinity chromatography | |
RUO | |
500235 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction