Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EMX1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA552294

Catalog No. PIPA552294


Only null left
Add to cart

Description

Description

Immunogen sequence: RSLYGGPELV FPEAMNHPAL TVHPAHQLGA SPLQPPHSFF GAQHRDPLHF YPWVLRNRFF GHRFQASDVP QDGLLLHGPF ARKPKRI Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 95%.

EMX1 is a transcription factor, which in cooperation with EMX2. It acts to generate the boundary between the roof and archipallium in the developing brain. It may function in combinations with OTX1/2 to specify cell fates in the developing central nervous system.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EMX1
Polyclonal
Unconjugated
Emx1
empty spiracles homeobox 1; empty spiracles homolog 1; Empty spiracles-like protein 1; EMX1; Emx-1; Homeobox protein EMX1; RGD1564002
Rabbit
Antigen affinity chromatography
RUO
13796, 2016
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.20 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q04741, Q04742
Emx1
Recombinant protein corresponding to Human EMX1.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.