Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ EN 219
GREENER_CHOICESDP

Catalog No. 74765 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-765 5 mg
74-762-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 74-765 Supplier Tocris Bioscience™ Supplier No. 7476/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Covalent RNF114 ligand

EN 219 is a covalent RNF114 inhibitor (IC50 = 470 nM). In vitro EN 219 mimics the mechanism of action of Nimbolide (Cat. No. 7289) by binding to RNF114 and inhibiting ubiquitination of its endogenous substrate, the tumor suppressor p21.

Specifications

Chemical Name or Material 1-[3,5-bis(4-Bromophenyl)-4,5-dihydro-1 H-pyrazol-1-yl]-2-chloroethanone
CAS 380351-29-1
Quantity 5 mg
Target RNF114
Molecular Formula C17H13Br2ClN2O
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.