Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EN-RAGE/S100A12 Antibody (133), PE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289816PE
Description
EN-RAGE/S100A12 Monoclonal antibody specifically detects EN-RAGE/S100A12 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
EN-RAGE/S100A12 | |
Monoclonal | |
PE | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
133 | |
Immunohistochemistry-Paraffin | |
CAAF1Neutrophil S100 protein, CAGCS100 calcium binding protein A12 (calgranulin C), Calcium-binding protein in amniotic fluid 1, Calgranulin-C, CGRPEN-RAGE, ENRAGE, Extracellular newly identified RAGE-binding protein, MRP6, p6calgranulin C, protein S100-A12, S100 calcium binding protein A12, S100 calcium-binding protein A12, S100 calcium-binding protein A12 (calgranulin C) | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human EN-RAGE/S100A12 (Accession#: NP_005612.1; Met1-Glu92). | |
0.1 mL | |
Cancer, Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
6283 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction