Learn More
Invitrogen™ EN1 Recombinant Rabbit Monoclonal Antibody (3B4K8)

Description
Positive Samples: SH-SY5Y, Mouse brain, Rat brain Immunogen sequence: KLKKKKNEKE DKRPRTAFTA EQLQRLKAEF QANRYITEQR RQTLAQELSL NESQIKIWFQ NKRAKIKKAT GIKNGLALHL MAQGLYNHST TTVQDKDESE.
Specifications
Specifications
| Antigen | EN1 |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 3B4K8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 |
| Gene | EN1 |
| Gene Accession No. | P09065 |
| Gene Alias | En1; En-1; engrailed 1; engrailed homeobox 1; engrailed homolog 1; engrailed-1; HME1; homeobox protein en-1; Homeobox protein engrailed-1; hu-En-1; Mo-en.1; mo-En-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.