Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EN2/Engrailed 2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981468
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1468 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1468 Supplier Bioss Supplier No. BS11552R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Homeobox-containing genes are thought to have a role in controlling development. In Drosophila, the ′engrailed′ (en) gene plays an important role during development in segmentation, where it is required for the formation of posterior compartments. Different mutations in the mouse homologs, En1 and En2, produced different developmental defects that frequently are lethal. The human engrailed homologs 1 and 2 encode homeodomain-containing proteins and have been implicated in the control of pattern formation during development of the central nervous system.

Specifications

Antigen EN2/Engrailed 2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene EN2
Gene Accession No. P19622
Gene Alias AUTS1; BB131122; EN2; En-2; engrailed 2; engrailed homeobox 2; engrailed homolog 2; engrailed-2; Homeobox protein en-2; Homeobox protein engrailed-2; hu-En-2; mo-En-2; RGD1561842
Gene Symbols EN2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human EN2/Engrailed 2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2020
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.