Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597706
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597706 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597706 Supplier Invitrogen™ Supplier No. PA597706
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EN2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Homeobox-containing genes are thought to have a role in controlling development. In Drosophila, the 'engrailed' (en) gene plays an important role during development in segmentation, where it is required for the formation of posterior compartments. Different mutations in the mouse homologs, En1 and En2, produced different developmental defects that frequently are lethal. The human engrailed homologs 1 and 2 encode homeodomain-containing proteins and have been implicated in the control of pattern formation during development of the central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EN2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene EN2
Gene Accession No. P19622
Gene Alias AUTS1; BB131122; EN2; En-2; engrailed 2; engrailed homeobox 2; engrailed homolog 2; engrailed-2; homeobox protein en-2; Homeobox protein engrailed-2; hu-En-2; mo-En-2; RGD1561842
Gene Symbols EN2
Host Species Rabbit
Immunogen Recombinant Human Homeobox protein engrailed-2 protein (1-251AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2020
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.