Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ EN6
GREENER_CHOICE

Catalog No. 747410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-741-0 10 mg
74-745-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 74-741-0 Supplier Tocris Bioscience™ Supplier No. 7474/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

H+ ATPase activator; activates autophagy; inactivates mTORC1 signaling

EN6 is an activator of lysosomal vacuolar H+ ATPase (v-ATPase). EN6 covalently binds to the catalytic subunit of v-ATPase leading to increased acidification of lysosomes, inactivation of mTORC1 signaling and activation of autophagy in vivo. EN6 triggers entosis and promotes clearance of TDP43 protein aggregates in vitro.

Specifications

Chemical Name or Material N-[4-Fluoro-3-[(1-oxo-2-propen-1-yl)amino]phenyl]-1-(2-fluorophenyl)-1 H-pyrazole-4-carboxamide
CAS 1808714-73-9
Quantity 10 mg
Target H+-ATPase Inhibitors
Molecular Formula C19H14F2N4O2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.