Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENAH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119841
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119841 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119841 Supplier Invitrogen™ Supplier No. PA5119841
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: SiHa cell lysate, 293T cell lysate, human breast carcinoma tissue, mouse testis tissue, SiHa. Subcellular Location: Cytoskeleton, cytoplasm, lamellipodium, filopodium, synapse, focal adhesion.

This gene encodes a member of the enabled/ vasodilator-stimulated phosphoprotein. Members of this gene family are involved in actin-based motility. This protein is involved in regulating the assembly of actin filaments and modulates cell adhesion and motility. Alternate splice variants of this gene have been correlated with tumor invasiveness in certain tissues and these variants may serve as prognostic markers. A pseudogene of this gene is found on chromosome 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENAH
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene ENAH
Gene Accession No. Q03173, Q8N8S7
Gene Alias ENA; enabled homolog; enabled homolog (Drosophila); ENAH; ENAH actin regulator; ENAH, actin regulator; mammalian enabled variant 11a; mammalian enabled variant pan; MENA; Ndpp1; NDPP-1; NPC derived proline rich protein 1; NPC-derived proline-rich protein 1; OTTHUMP00000035864; Protein enabled homolog; WBP8
Gene Symbols ENAH
Host Species Rabbit
Immunogen Synthetic peptide within human ENAH aa 100-150.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13800, 55740
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.