Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENC-2 Monoclonal Antibody (1B7)
GREENER_CHOICE

Catalog No. PIMA515656
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515656 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515656 Supplier Invitrogen™ Supplier No. MA515656
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15656 targets KLHL25 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15656 immunogen is purified recombinant fragment of human KLHL25 expressed in E. Coli. . MA5-15656 detects KLHL25 which has a predicted molecular weight of approximately 65kDa.

KLHL25 (ectoderm-neural cortex protein 2, ENC2) is a cytoplasmic protein that contains six Kelch regions and a single BTB (POZ) domain. KLHL25 is highly homologus to another Kelch-like protein, ENC1, and it is believed to operate in a manner similar to other Kelch-domain containing proteins. Kelch-domain repeat containing proteins often act as modifiers of Actin fibers. Expressed early in embryogenesis, ENC1 helps to mediate neuronal process formation. It also appears to have a role in neural crest cell differentiation. KLHL25 likely functions as a substrate specific adapter for protein ubiquitinating complexes. KLHL25 is expressed in most tissues with highest expression in brain and liver.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENC-2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1B7
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene KLHL25
Gene Accession No. Q9H0H3
Gene Alias 2810402K13Rik; BC027373; BTB/POZ KELCH domain protein; ectodermal-neural cortex 2; ectoderm-neural cortex protein 2; ectoderm-neural cortex-1; Enc-1; ENC2; ENC-2; FLJ12587; FLJ45015; kelch like family member 25; kelch-like 25; kelch-like family member 25; kelch-like protein 25; KLHL25; RGD1310815
Gene Symbols KLHL25
Host Species Mouse
Immunogen Purified recombinant fragment of human KLHL25 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64410
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.