Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENC-2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582832
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582832 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582832 Supplier Invitrogen™ Supplier No. PA582832
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HFETVRQSED FNSLSKDTLL DLISSDELET EDERVVFEAI LQWVKHDLEP RKVHLPELLR SVRLALLPSD CLQEAVSSEA LLMADERTKL IMDEALRCKT RILQNDGVVT SPCA Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 88%.

KLHL25 (ectoderm-neural cortex protein 2, ENC2) is a cytoplasmic protein that contains six Kelch regions and a single BTB (POZ) domain. KLHL25 is highly homologus to another Kelch-like protein, ENC1, and it is believed to operate in a manner similar to other Kelch-domain containing proteins. Kelch-domain repeat containing proteins often act as modifiers of Actin fibers. Expressed early in embryogenesis, ENC1 helps to mediate neuronal process formation. It also appears to have a role in neural crest cell differentiation. KLHL25 likely functions as a substrate specific adapter for protein ubiquitinating complexes. KLHL25 is expressed in most tissues with highest expression in brain and liver.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENC-2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KLHL25
Gene Accession No. Q9H0H3
Gene Alias 2810402K13Rik; BC027373; BTB/POZ KELCH domain protein; ectodermal-neural cortex 2; ectoderm-neural cortex protein 2; ectoderm-neural cortex-1; Enc-1; ENC2; ENC-2; FLJ12587; FLJ45015; kelch like family member 25; kelch-like 25; kelch-like family member 25; kelch-like protein 25; KLHL25; RGD1310815
Gene Symbols KLHL25
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KLHL25. Recombinant protein control fragment (Product #RP-93633).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64410
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.