Learn More
Invitrogen™ ENC-2 Polyclonal Antibody

Description
A suggested positive control is Jurkat cell lysate. PA5-20937 can be used with blocking peptide PEP-1051.
Specifications
Specifications
| Antigen | ENC-2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | KLHL25 |
| Gene Accession No. | Q9H0H3 |
| Gene Alias | 2810402K13Rik; BC027373; BTB/POZ KELCH domain protein; ectodermal-neural cortex 2; ectoderm-neural cortex protein 2; ectoderm-neural cortex-1; Enc-1; ENC2; ENC-2; FLJ12587; FLJ45015; kelch like family member 25; kelch-like 25; kelch-like family member 25; kelch-like protein 25; KLHL25; RGD1310815 |
| Gene Symbols | KLHL25 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.