Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103088
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103088 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103088 Supplier Invitrogen™ Supplier No. PA5103088
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ENC1.

The ectoderm-neural cortex-1 (ENC-1) protein is an early and highly specific marker of neural induction in vertebrates. It is a kelch family related protein that functions as an actin-binding protein and has been suggested to be involved in the organization of the actin cytoskeleton during neural fate specification and development of the nervous system. ENC-1 has also been shown to be required for adipocyte differentiation when cytoskeletal reorganization and cell shape change from fibroblastic preadipocytes to spherical adipocytes occur.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENC1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ENC1
Gene Accession No. O14682, O35709
Gene Alias CCL28; ectodermal-neural cortex (with BTB-like domain); ectodermal-neural cortex 1; ectodermal-neural cortex 1 (with BTB domain); ectodermal-neural cortex 1 (with BTB-like domain); Ectoderm-neural cortex protein 1; Enc1; ENC-1; FLJ39259; kelch-like 35; kelch-like 37; kelch-like family member 37; kelch-like protein 37; KLHL35; KLHL37; NRPB; Nuclear matrix protein NRP/B; nuclear restrict protein in brain; nuclear restricted protein, BTB domain-like (brain); p53-induced gene 10 protein; PIG10; si:dkey-35i22.1; TP53I10; tumor protein p53 inducible protein 10
Gene Symbols ENC1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ENC1(Accession O14682), corresponding to amino acid residues C59-R109.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13803, 294674, 8507
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.