Learn More
Sigma Aldrich EncapGel-UV Kit, photo-crosslinkable sIPN hydrogel kit
Catalog No. 7996101KT Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 kt
Catalog No. 79-961-01KT
Supplier: Sigma Aldrich 7996101KT
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
EncapGel(TM)-UV Kit is a ready-to-use, UV- crosslinked hydrogel kit for the encapsulation and delivery of drugs, biomolecules, and cells. This hydrogel fuses the durability of synthetic polymers and the biorecognition of natural proteins to create a biocompatible delivery platform. The biodegradable matrix allows for temporally and spacially controlled delivery. Applications include:Localized delivery of small molecules, proteins, nucleic acids, and cellular-based therapeutics Tissue engineeringRegenerative medicineENCAPGEL is a trademark of Sigma-Aldrich Co. LLC
Specifications
Quantity 1 kt
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.