Learn More
Ansell ENCORE™ Latex Textured Surgical Gloves
GREENER_CHOICE
Catalog No. 19501144 Shop All Ansell Products
Change view
Click to view available options
Size:
5.5
6
6.5
7
7.5
8
8.5
9
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-501-144 6
19-501-143 5.5
19-501-145 6.5
19-501-146 7
19-501-147 7.5
19-151-226 8
19-501-149 8.5
19-501-150 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-501-144 Supplier Ansell Supplier No. 5785001
Add to Cart
Add to Cart
This item is not returnable. View return policy
Latex, powder free surgical gloves with a textured surface for exceptional instrument handling and grip.
  • Natural rubber latex
  • Textured finish ensures secure grip under dry and damp conditions
  • Ideally used as outer glove when double-gloving
  • Optimal tactile sensitivity and protection
  • DERMASHIELD™ Technology internal coating facilitates intraoperative changes
  • Effortless dry and damp donning
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ASTM D3577, ASTM D6978-05, ASTM D7160, ASTM F739
Certifications/Compliance (Europe) EN 16523-1, EN 374:2003, EN 420:2003 + A1:2009, EN 455 1-4, EN ISO 374-1:2016, EN ISO 374-5:2016, ISO 10282
AQL 0.65
Color Natural
Exterior Finish Fully Textured
Laboratory/Cleanroom Use No
Material Latex
Size 6
Sterility Sterile
Manufacturing/QMS Audit Standards EN 556, ISO 11137-Part 1, ISO 13485, ISO 14001, ISO 9001
Model 5785001
Series ENCORE™ Latex Textured
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.