Learn More
Endfitting with Fixed 1 μm Frit for TSKgel™ Super Series RP and SuperH2 Columns, Sold by MilliporeSigma™ Supelco™
Catalog No. 111022352 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Particle Size:
1 μm
Catalog No. Particle Size
11-102-2352 1 μm
1 options
Catalog No. 11-102-2352 Supplier MilliporeSigma Supelco Supplier No. 818255
Add to Cart
Add to Cart
Replacement endfitting for maintaining optimal column performance in TSKgel® HPLC applications.
  • Material: Stainless steel frit
  • Pore size: 1 μm
  • Compatibility: TSKgel Super Series RP and SuperH2 columns
  • Techniques: HPLC
  • Packaging: 1 ea per package
TRUSTED_SUSTAINABILITY
Specifications
For Use With TSKgel™ Super Series Columns
Particle Size 1 μm
Pore Size Not Applicable
Quantity 1 Ea.
Product Line TSKgel
Material Stainless Steel
Technique HPLC
Product Type Endfitting
TSKgel is a registered trademark of Tosoh Corporation