Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endo G Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 2 publications

Supplier:  Novus Biologicals NBP176657

Catalog No. NBP176657


Only null left
Add to Cart

Description

Description

Endo G Polyclonal specifically detects Endo G in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications

Specifications

Endo G
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
EC 3.1.30.-, Endo G, endonuclease G, endonuclease G, mitochondrial, FLJ27463, mitochondrial endonuclease G
Rabbit
33 kDa
0.1 mg
Apoptosis, Cancer, Hypoxia
2021
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
1 mg/mL
Western Blot 0.5-2 ug/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5-15 ug/mL, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin, Immunofluorescence 20 ug/ml
Q14249
ENDOG
Antibody was raised with a synthetic peptide corresponding to 15 amino acids near the amino terminus of human EndoG. The immunogen is located within amino acids 40 - 90 of EndoG. Amino Acid Squence: GGPRGPGELAKYGLP
Affinity Purified
RUO
Primary
Human, Mouse, Rat, Canine
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only