Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Endo G Antibody (JU31-30), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP275480
Description
Endo G Monoclonal specifically detects Endo G in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Endo G | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
EC 3.1.30.-, Endo G, endonuclease G, endonuclease G, mitochondrial, FLJ27463, mitochondrial endonuclease G | |
Rabbit | |
36 kDa | |
100 μL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
JU31-30 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
ENDOG | |
Synthetic peptide within Human Endo G aa 200-297/ 297. (SwissProt: Q14249 Human) | |
Protein A purified | |
Apoptosis, Cancer, Hypoxia | |
2021.0 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction