Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endocan Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579221
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579221 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579221 Supplier Thermo Scientific Supplier No. PA579221
Add to Cart
Add to Cart

Endocan Polyclonal Antibody

Endocan Polyclonal Antibody for Western Blot, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Endocan
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene ESM1
Gene Accession No. Q9NQ30
Gene Alias 0610042H23Rik; AV004503; endocan; endothelial cell specific molecule 1; endothelial cell-specific molecule 1; ESM1; ESM-1; ESM-1 secretory protein; IGFBP-rp6; PG25; pineal specific PG25 protein
Gene Symbols ESM1
Host Species Rabbit
Immunogen E. coli-derived human ESM1 recombinant protein (Position: W20-R184).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11082
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.