Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Endocan Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144096
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144096 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144096 Supplier Invitrogen™ Supplier No. PA5144096
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Endocan Polyclonal Antibody for IHC (P), ELISA

Endothelial cell-specific molecule 1 is a protein that in humans is encoded by the ESM1 gene. This gene encodes a secreted protein which is mainly expressed in the endothelial cells in human lung and kidney tissues. The expression of this gene is regulated by cytokines, suggesting that it may play a role in endothelium-dependent pathological disorders. The transcript contains multiple polyadenylation and mRNA instability signals. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Endocan
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene ESM1
Gene Accession No. Q9NQ30
Gene Alias 0610042H23Rik; AV004503; endocan; endothelial cell specific molecule 1; endothelial cell-specific molecule 1; ESM1; ESM-1; ESM-1 secretory protein; IGFBP-rp6; PG25; pineal specific PG25 protein
Gene Symbols ESM1
Host Species Rabbit
Immunogen Recombinant Human Endothelial cell-specific molecule 1 protein (20-184AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11082
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.