Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENDOD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552701
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552701 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552701 Supplier Invitrogen™ Supplier No. PA552701
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GECDKFFYAG TPPAGLAADS HVKICQRAEG AERFATLYST RDRIPVYSAF RAPRPAPGGA EQRWLVEPQI DDPNSNLEEA INEAEAITSV NSLGSKQALN TDYLDSDYQR GQLYPFSLSS DVQVATFTLT NSAPMTQSF Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 21%.

May act as a DNase and a RNase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENDOD1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ENDOD1
Gene Accession No. O94919
Gene Alias 2210414F18Rik; 2310067E08Rik; C85344; Endod1; endonuclease domain containing 1; endonuclease domain-containing 1 protein; Kiaa0830; RGD1310444
Gene Symbols ENDOD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ENDOD1. Recombinant protein control fragment (Product #RP-89925).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23052
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.