Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ENDOD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568187
Description
Predicted to react with Mouse samples.
May act as a DNase and a RNase.
Specifications
ENDOD1 | |
Polyclonal | |
Unconjugated | |
ENDOD1 | |
2210414F18Rik; 2310067E08Rik; C85344; Endod1; endonuclease domain containing 1; endonuclease domain-containing 1 protein; Kiaa0830; RGD1310444 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23052, 71946 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O94919, Q8C522 | |
ENDOD1 | |
Synthesized peptide derived from human ENDOD1. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction