Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endofin Polyclonal Antibody, Huabio

Catalog No. 502544574
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4574 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4574 Supplier HUABIO RESEARCH INC Supplier No. HA500231
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Endofin Polyclonal antibody specifically detects Endofin in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Endofin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene ZFYVE16
Gene Accession No. Q7Z3T8, Q80U44
Gene Alias AI035632; B130024H06Rik; B130031L15; Endofin; endosomal-associated FYVE domain protein; Endosome-associated FYVE domain protein; endosome-associated FYVE-domain protein; Kiaa0305; mKIAA0305; PPP1R69; protein phosphatase 1, regulatory subunit 69; RGD1564784; ZFYVE16; Zinc finger FYVE domain-containing protein 16; zinc finger FYVE-type containing 16; zinc finger, FYVE domain containing 16
Host Species Rabbit
Immunogen Recombinant protein within human Endofin aa 1-200.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 218441, 9765
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.