Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENDOG Monoclonal Antibody (7F2G10)
GREENER_CHOICE

Catalog No. PIMA515759
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515759 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515759 Supplier Invitrogen™ Supplier No. MA515759
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

A suggested positive control is HepG2 cell lysate.

The protein encoded by this gene is a nuclear encoded endonuclease that is localized in the mitochondrion. The encoded protein is widely distributed among animals and cleaves DNA at GC tracts. This protein is capable of generating the RNA primers required by DNA polymerase gamma to initiate replication of mitochondrial DNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENDOG
Applications Western Blot
Classification Monoclonal
Clone 7F2G10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ENDOG
Gene Accession No. Q14249
Gene Alias endo G; Endog; endonuclease G; endonuclease G, mitochondrial; mitochondrial endonuclease G
Gene Symbols ENDOG
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 51 - 140 of human EndoG.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2021, 362100
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.