Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENDOG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587462
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587462 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587462 Supplier Invitrogen™ Supplier No. PA587462
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AELPPVPGGP RGPGELAKYG LPGLAQLKSR ESYVLCYDPR TRGALWVVEQ LRPERLRGDG DRRECDFRED DSVHAYHRAT NADYRGSGFD RGHLAAAANH RWSQKAMDDT FYLSNVAPQV PHLNQNAWNN LEKYSRSLTR SYQNVYVCTG PLFLPRTEAD GKSYVKYQVI GKNHVAVPTH FFKVLILEAA GGQIELRTYV MPNAPVDEAI PLERFLVPIE SIERASGLLF VPNILARAGS LKAITAGSK; Positive Samples: HeLa, MCF7, SW620, Mouse heart, Mouse kidney, Rat heart; Cellular Location: Mitochondrion.

The protein encoded by this gene is a nuclear encoded endonuclease that is localized in the mitochondrion. The encoded protein is widely distributed among animals and cleaves DNA at GC tracts. This protein is capable of generating the RNA primers required by DNA polymerase gamma to initiate replication of mitochondrial DNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENDOG
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.99 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ENDOG
Gene Accession No. O08600, Q14249
Gene Alias endo G; Endog; endonuclease G; endonuclease G, mitochondrial; mitochondrial endonuclease G
Gene Symbols ENDOG
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 49-297 of human Endo G (NP_0044262).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13804, 2021, 362100
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.