Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENDOGL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143251
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143251 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5143251 Supplier Invitrogen™ Supplier No. PA5143251
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize both reported isoforms (NP_005098.2; NP_001138936.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

This gene encodes an endo/exonuclease with 5'-3' exonuclease activity. The encoded enzyme catalyzes the hydrolysis of ester linkages at the 5' end of a nucleic acid chain. This enzyme is localized to the mitochondria and may play a role in programmed cell death. Alternatively spliced transcript variants have been described. A pseudogene exists on chromosome 18.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENDOGL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene EXOG
Gene Accession No. Q9Y2C4
Gene Alias AW557704; Endo G-like 1; endo/exonuclease (5'-3'), endonuclease G-like; endo/exonuclease (5-3), endonuclease G-like; ENDOGL1; Endogl2; endonuclease G-like 1; endonuclease G-like 2; ENGL; Engla; ENGL-a; ENGLB; ENGL-b; exo/endonuclease G; EXOG; Nuclease EXOG, mitochondrial; nuclease EXOG, mitochondrial-like; Nuclease EXOG, mitochondrial-like protein
Gene Symbols EXOG
Host Species Goat
Immunogen Peptide with sequence C-KAKEQSGTQIRKP (aa 355-367).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9941
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.