Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Endoglin/CD105 Antibody (103), FITC

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289299F
Description
Endoglin/CD105 Monoclonal specifically detects Endoglin/CD105 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
Endoglin/CD105 | |
Monoclonal | |
FITC | |
CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Endoglin/CD105 (NP_001108225.1; Met1-Gly586). | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
2022 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry, Immunofluorescence, Immunocytochemistry | |
103 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction