Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Endoglin/CD105 Antibody (MJ7/18), Alexa Fluor™ 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281085AF594
Description
Endoglin/CD105 Monoclonal specifically detects Endoglin/CD105 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Endoglin/CD105 | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
ENG | |
MJ7/18 was prepared by immunizing rats with inflamed mouse skin tissue. The antibody was selected immunohistologically for reactivity with endothelial cells. | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
2022 | |
Mouse | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
MJ7/18 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1 | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
MJ7/18 binds specifically to mouse Endoglin/CD105. Endoglin/CD105 is a homodimer expressed by vascular endothelial cells. Endoglin/CD105 functions in adhesion and embryonic angiogenesis. Endoglin/CD105 is a major homodimeric glycoprotein of vascular endothelium involved in cell ahesion through integrin/RGD binding and the regulation of angiogenesis. Also acts as TGF-beta coreceptor and is involved in the TGF-beta/BMP signaling cascade. Endoglin/CD105 is also required for GDF2/BMP9 signaling through SMAD1 in endothelial cells. | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction