Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endoglin/CD105 Antibody (SN6) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$192.47 - $529.95

Specifications

Antigen Endoglin/CD105
Clone SN6
Concentration 1 mg/mL
Dilution Western Blot 1:100 - 1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500
Applications Immunohistochemistry (Frozen), Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB017734
SDP
View Documents
Novus Biologicals
NB100656010.025MG
0.025 mg
Each for $192.47
Only null left
Add to Cart
 
NB10065601
SDP
View Documents
Novus Biologicals
NB10065601
0.1 mg
Each for $529.95
Only null left
Add to Cart
 
Description

Description

Endoglin/CD105 Monoclonal specifically detects Endoglin/CD105 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications

Specifications

Endoglin/CD105
1 mg/mL
Immunohistochemistry (Frozen), Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
Mouse
Cancer, Cell Biology, Endothelial Cell Markers, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers
P17813
2022
Partially purified cell membrane antigens from fresh leukemia cells.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
SN6
Western Blot 1:100 - 1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500
Monoclonal
Purified
RUO
Human
CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1
ENG
IgG1
Protein G purified
Recognizes the human CD105 cell surface antigen, a glycoprotein homodimer of 95kD subunits. CD105 is also known as endoglin, and is expressed by endothelial cells, activated monocytes and some leukemia cells.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.