Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Endoglin/CD105 Antibody (SN6), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065601MFV610
Description
Endoglin/CD105 Monoclonal antibody specifically detects Endoglin/CD105 in Human samples. It is validated for Flow CytometrySpecifications
Endoglin/CD105 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
SN6 | |
Flow Cytometry | |
CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1 | |
Partially purified cell membrane antigens from fresh leukemia cells. | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
2022 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction