Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endoglin/CD105 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735816 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5816 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5816 Supplier Proteintech Group Inc Supplier No. 281171AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Endoglin (Osler-Rendu-Weber syndrome 1), CD105, is a type I membrane glycoprotein located on cell surfaces and is a part of the TGF beta receptor complex. Its gene is mapped to human chromosome 8. The protein consists of a homodimer of 180 kDA with disulfide links. It has been found on endothelial cells, activated macrophages, fibroblasts and smooth muscle cells. Endoglin has a role in the development of the cardiovascular system and in vascular remodeling and has been found to be elevated in pregt women who subsequently develop preeclampsia.

Specifications

Antigen Endoglin/CD105
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ENG
Gene Accession No. P17813
Gene Alias CD105, END, Endoglin, ENG, FLJ41744, HHT1, ORW, ORW1
Gene Symbols ENG
Host Species Rabbit
Immunogen Endoglin/CD105 Fusion Protein Ag27822
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2022
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.