Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Endoglycan/PODXL2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Endoglycan/PODXL2 |
---|---|
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Endoglycan/PODXL2 Polyclonal specifically detects Endoglycan/PODXL2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Endoglycan/PODXL2 | |
Polyclonal | |
Rabbit | |
Human | |
Q9NZ53 | |
50512 | |
This antibody was developed against a recombinant protein corresponding to amino acids: EAQSRIPWDSTQVICKDWSNLAGKNYIILNMTENIDCEVFRQHRGPQLLALVEEVLPRHGSGHHGAWHISLSKPSEKEQHLLMTLVGEQGVVPTQDVLSMLGDIRRSLEEIGIQNYSTTSS | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
endoglycan, PODLX2, podocalyxin-like 2, podocalyxin-like protein 2 | |
PODXL2 | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title