Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EndoGlyx-1 Monoclonal Antibody (H572), eBioscience™
GREENER_CHOICE

Catalog No. 5017838
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-178-38 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-178-38 Supplier Invitrogen™ Supplier No. BMS169
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: EndoGlyx-1 is a pan-endothelial human cell surface glycoprotein complex composed of four different disulfide-bonded protein species with an apparent molecular mass of approximately 500 kDa. BMS169 specifically reacts the glycoprotein EndoGlyx-1 that is expressed on endothelial cells of human normal and tumor blood vessel, but not on cells of mesenchymal, neuroectodermal and epithelial derivation. Applications Tested: Flow Cytometry, Immunohistochemistry, Immunoprecipitation.

Inhibits endothelial cells motility and acts as a negative regulator of angiogenesis; it downregulates KDR activation by binding VEGFA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EndoGlyx-1
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone H572
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene MMRN2
Gene Accession No. Q9H8L6
Gene Alias Elastin microfibril interface located protein 3; elastin microfibril interfacer 3; EMILIN3; EMILIN-3; EMILIN-like protein EndoGlyx-1; ENDOGLYX-1; endoGlyx-1 p125/p140 subunit; MMRN2; multimerin 2; multimerin-2
Gene Symbols MMRN2
Host Species Mouse
Immunogen Rec. Human EndoGlyx-1.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79812
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.