Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EndoGlyx-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554264
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554264 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554264 Supplier Invitrogen™ Supplier No. PA554264
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HAQHFTLHRS ISELQADVDT KLKRLHKAQE APGTNGSLVL ATPGAGARPE PDSLQARLGQ LQRNLSELHM TTARREEELQ YTLEDMRATL TRHVDEIKEL YSESDETFDQ ISKVERQVEE LQVNHTALRE LRVILMEK Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 73%.

Inhibits endothelial cells motility and acts as a negative regulator of angiogenesis; it downregulates KDR activation by binding VEGFA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EndoGlyx-1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MMRN2
Gene Accession No. Q9H8L6
Gene Alias Elastin microfibril interface located protein 3; elastin microfibril interfacer 3; EMILIN3; EMILIN-3; EMILIN-like protein EndoGlyx-1; ENDOGLYX-1; endoGlyx-1 p125/p140 subunit; MMRN2; multimerin 2; multimerin-2
Gene Symbols MMRN2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EndoGlyx-1. Recombinant protein control fragment (Product #RP-102444).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79812
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.