Learn More
Invitrogen™ Endomucin Monoclonal Antibody (eBioV.7C7 (V.7C7)), eFluor™ 660, eBioscience™, Invitrogen™

Description
Description: The eBioV.7C7 monoclonal antibody reacts with mouse endomucin, which was identified in a search for cell-surface expressed endothelial cell markers. Endomucin is a 75 kDa type I integral membrane protein, with similarities to the sialomucin family of proteins including extensive O-linked glycosylation. Endomucin is expressed on endothelial cells, however, an exception is the high endothelial venules (HEV) of secondary lymphoid organs. In addition, it has been demonstrated that endomucin is expressed on CD34-c-Kit+Sca-1+Lin- hematopoietic progenitors, and that these cells are capable of multi-lineage long-term reconstitution of the hematopoietic compartment. Applications Reported: This eBioV.7C7 (V.7C7) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioV.7C7 (V.7C7) antibody has been tested by flow cytometric analysis of bEnd.3 cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm).
Specifications
Specifications
| Antigen | Endomucin |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | eBioV.7C7 (V.7C7) |
| Concentration | 0.2 mg/mL |
| Conjugate | eFluor 660 |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | Emcn |
| Gene Accession No. | Q9R0H2 |
| Gene Alias | 0610012K22Rik; AI315669; Emcn; EMCN2; Endomucin; endomucin-1; endomucin-1/2; Endomucin-2; gastric cancer antigen Ga34; Muc14; MUC-14; mucin 14, endothelial; mucin-14; sialomucin |
| Show More |
Frequently Asked Questions (FAQs)
Our options will depend on the samples you are analyzing.
If cell viability is not critical, you can store your stained samples at 4 degrees C or on ice overnight in the dark and analyze the following day.
For samples stained with eFluor organic fluorochromes, we recommend that cells be suspended in 100 uL of Flow Cytometry Staining Buffer (Cat. No. 00-4222) and 100 uL of eBioscience IC Fixation Buffer (Cat. No. 00-8222); samples can be incubated for up to 3 days at 4 degrees C in the dark. Alternatively, the 1-step Fix/Lyse Solution (Cat. No. 00-5333) can be used. This is a great option when working with whole blood but also works for other cell types.
Yes, the eFluor Organic fluorochromes can be used for intracellular staining. The eFluor organic fluorochromes maintain bright signal and require minimal changes in compensation when fixed with eBioscience IC Fixation Buffer (Cat. No. 00-8222-49) and Permeabilization Buffer (Cat. No. 00-8333-56) or 1-step Fix/Lyse Solution (Cat. No. 00-5333-54, 00-5333-57) (as compared to live cells).
Yes, in-house studies have demonstrated that the eFluor 660 fluorochrome is recognized by Anti-Cy5/Alexa Fluor 647 beads. Side by side studies with Alexa Fluor 647 versus eFluor 660 conjugated antibodies have demonstrated comparable results.
The eFluor Organic fluorochromes and eVolve QDots can be used with flow staining buffers containing PBS and protein.
The eFluor Organic Dyes (eFluor 450, APC-eFluor 780, PerCP-eFluor 710, eFluor 710) are conventional fluorochromes. In contrast, the eVolve line of products are Quantum dots.
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.