Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Endomucin/Mucin -14 Antibody [Janelia Fluor™ 525], Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals AF4666JF525
Description
Endomucin/Mucin -14 Polyclonal antibody specifically detects Endomucin/Mucin -14 in Mouse samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry,CyTOFSpecifications
Endomucin/Mucin -14 | |
Polyclonal | |
50mM Sodium Borate | |
Goat | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, Western Blot, Immunocytochemistry, CyTOF | |
Janelia Fluor 525 | |
endomucin, MUC-14 | |
E. coli-derived recombinant mouse Endomucin, Glu21-Glu90, Accession # Q9R0H2 | |
0.1 mL | |
Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
51705 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction