Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Endomucin Recombinant Rabbit Monoclonal Antibody (8Y9J9)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551480
Description
Endomucin Recombinant Monoclonal Antibody for IHC (P), ELISA
Endomucin (EMCN) also known as endothelial sialomucin or mucin-like sialoglycoprotein interferes with the assembly of focal adhesion complexes and inhibits interaction between cells and the extracellular matrix.
Specifications
Endomucin | |
Recombinant Monoclonal | |
0.26 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q9R0H2 | |
Emcn | |
Recombinant protein of human Endomucin. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
8Y9J9 | |
Unconjugated | |
Emcn | |
0610012K22Rik; AI315669; Emcn; EMCN2; Endomucin; endomucin-1; endomucin-1/2; Endomucin-2; gastric cancer antigen Ga34; Muc14; MUC-14; mucin 14, endothelial; mucin-14; sialomucin | |
Rabbit | |
Affinity chromatography | |
RUO | |
59308 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction