Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Endophilin B1/B2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7456SP
This item is not returnable.
View return policy
Description
Endophilin B1/B2 Monoclonal specifically detects Endophilin B1/B2 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.Specifications
Endophilin B1/B2 | |
Monoclonal | |
LYOPH | |
Western Blot 0.2 ug/mL, Simple Western 2 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 5-25 ug/mL | |
Q9Y371 | |
E. coli-derived recombinant human Endophilin B1/Bif-1 Ala33-Asn189 Accession # Q9Y371 | |
25 μg | |
Primary | |
Detects human Endophilin B1/Bif-1 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human Endophilin B2 is observed. | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
807009 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
51100 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction