Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Endosialin/CD248/TEM1 Antibody (OTI4B8), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272075AF532
Description
Endosialin/CD248/TEM1 Monoclonal specifically detects Endosialin/CD248/TEM1 in Human samples. It is validated for Western Blot.Specifications
Endosialin/CD248/TEM1 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
CD248 | |
Recombinant protein fragment corresponding to amino acids 20-300 of human Endosialin/CD248/TEM1 (NP_065137) produced in E.coli. | |
Primary | |
Human | |
IgG2b |
Western Blot | |
OTI4B8 | |
Western Blot | |
2610111G01Rik, CD164 sialomucin-like 1, CD164L1, CD248 antigen, CD248 antigen, endosialin, CD248 molecule, endosialin, endosialin, MGC119479, TEM1MGC119478, Tumor endothelial marker 1 | |
Mouse | |
0.1 mL | |
57124 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction