Learn More
Endostatin Antibody (RM0060-3H11), Alexa Fluor™ 405, Novus Biologicals™
SDP
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NBP212274AF405
Catalog No. NB212274405
Only null left
Add to cart
Description
Description
Endostatin Monoclonal specifically detects Endostatin in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Endostatin
Monoclonal
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
COL18A1
Mouse recombinant Endostatin
0.1 mL
Primary
Mouse
Purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
RM0060-3H11
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
antiangiogenic agent, collagen alpha-1(XVIII) chain, collagen, type XVIII, alpha 1, FLJ27325, FLJ34914, human type XVIII collagen10endostatin, KNO, KNO1MGC74745, Knobloch syndrome, type 1, KS, multi-functional protein MFP
Rat
Protein A or G purified
Angiogenesis, Biologically Active Proteins, Cancer, Extracellular Matrix, Tumor Suppressors
80781
Store at 4C in the dark.
IgG2
Product Suggestions
Product Suggestions
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.