Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Endostatin Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP31765525UL
This item is not returnable.
View return policy
Description
Endostatin Polyclonal antibody specifically detects Endostatin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Endostatin | |
Polyclonal | |
Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
antiangiogenic agent, collagen alpha-1(XVIII) chain, collagen, type XVIII, alpha 1, FLJ27325, FLJ34914, human type XVIII collagen10endostatin, KNO, KNO1MGC74745, Knobloch syndrome, type 1, KS, multi-functional protein MFP | |
This antibody was developed against Recombinant Protein corresponding to amino acids: ADPDKFQGVIAELKVRRDPQVSPMHCLDEEGDDSDGASGDSGSGLGDARELLREETGAALKPRLPAPPPVTTPPLAGGSSTEDSRSEEVE | |
25 μg | |
Angiogenesis, Biologically Active Proteins, Cancer, Extracellular Matrix, Tumor Suppressors | |
80781 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction