Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Endothelial Lipase Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB400118AF594
Description
Endothelial Lipase Polyclonal specifically detects Endothelial Lipase in Human, Mouse (Negative) samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Endothelial Lipase | |
Polyclonal | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
EC 3.1.1, EC 3.1.1.3, EDLendothelial lipase, ELlipoprotein lipase H, Endothelial cell-derived lipase, lipase, endothelial, PRO719 | |
Rabbit | |
0.1 mL | |
9388 | |
Store at 4°C in the dark. |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
LIPG | |
An N-terminal synthetic peptide made to human Endothelial Lipase [UniProtNo. Q9Y5X9] | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction