Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Endothelial Lipase Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB400118H
Description
Endothelial Lipase Polyclonal specifically detects Endothelial Lipase in Human, Mouse (Negative) samples. It is validated for Western Blot, ELISA.Specifications
Endothelial Lipase | |
Polyclonal | |
Western Blot, ELISA | |
Q9Y5X9 | |
LIPG | |
An N-terminal synthetic peptide made to human Endothelial Lipase [UniProt# Q9Y5X9] | |
0.1 mL | |
Autophagy, Endothelial Cell Markers, Lipid and Metabolism | |
9388 | |
Human, Mouse (Negative) | |
IgG |
Western Blot, ELISA | |
HRP | |
PBS with No Preservative | |
EC 3.1.1, EC 3.1.1.3, EDLendothelial lipase, ELlipoprotein lipase H, Endothelial cell-derived lipase, lipase, endothelial, PRO719 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human. Does not appear to recognize mouse. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction