Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Endothelial Lipase Antibody (OTI1D9), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP202396
Description
Endothelial Lipase Monoclonal specifically detects Endothelial Lipase in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Endothelial Lipase | |
Monoclonal | |
0.95 mg/mL | |
Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 2ug/500ul, Immunohistochemistry-Paraffin 1:50 | |
LIPG | |
Full length human recombinant protein of human LIPG (NP_006024) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG2b |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
OTI1D9 | |
Unconjugated | |
EC 3.1.1, EC 3.1.1.3, EDLendothelial lipase, ELlipoprotein lipase H, Endothelial cell-derived lipase, lipase, endothelial, PRO719 | |
Mouse | |
54.6 kDa | |
0.1 mL | |
Autophagy, Cholesterol Metabolism, Endothelial Cell Markers, Lipid and Metabolism, Unfolded Protein Response | |
9388 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction