Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Endothelin 1 Monoclonal Antibody (3G10)
GREENER_CHOICE

Catalog No. MA183134
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA183134 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA183134 Supplier Invitrogen™ Supplier No. MA183134
Add to Cart
Add to Cart

Rat Monoclonal Antibody

For IHC applications, frozen sections processed with dry acetone are recommended. Paraformaldehyde is not recommended as it destroys the epitope. Rat anti Human endothelin antibody, clone 3G10 reacts with the C terminal of endothelins 1, 2 and 3.

The protein encoded by this gene is proteolytically processed to release a secreted peptide termed endothelin 1. This peptide is a potent vasoconstrictor and is produced by vascular endothelial cells. Endothelin 1 also can affect the central nervous system. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Endothelin 1
Applications Immunohistochemistry (Frozen), Immunoradiometric Assay
Classification Monoclonal
Clone 3G10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene EDN1
Gene Accession No. P05305
Gene Alias ARCND3; Big endothelin 1; Big endothelin-1; Big ET; Big ET 1; Big ET1; BigET; EDN1; endothelin 1; endothelin-1; Et; ET1; ET-1; HDLCQ7; PPET1; ppET-1; preproendothelin; preproendothelin-1; preproET; QME
Gene Symbols EDN1
Host Species Rat
Immunogen Synthetic peptide corresponding to the c-terminal region of human endothelin (C,H,L,D,I,I,W), conjugated to BSA.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1906
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.