Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endothelin-1 Mouse anti-Human, Clone: 2C4, Bioss
SDP

Catalog No. 501979876
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-197-9876 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-197-9876 Supplier Bioss Supplier No. BSM0954M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The protein encoded by this gene is proteolytically processed to release a secreted peptide termed endothelin 1. This peptide is a potent vasoconstrictor and is produced by vascular endothelial cells. Endothelin 1 also can affect the central nervous system. Two transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen Endothelin-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2C4
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene EDN1
Gene Accession No. P05305
Gene Alias ARCND3; Big endothelin 1; Big endothelin-1; Big ET; Big ET 1; Big ET1; BigET; EDN1; endothelin 1; Endothelin-1; Et; ET1; ET-1; HDLCQ7; PPET1; PPET-1; preproendothelin; Preproendothelin-1; preproET; QME
Gene Symbols EDN1
Host Species Mouse
Immunogen KLH conjugated synthetic peptide derived from human Endothelin-1.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1906
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.