Learn More
Invitrogen™ Endothelin 1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587481
Description
Immunogen sequence: APETAVLGAE LSAVGENGGE KPTPSPPWRL RRSKRCSCSS LMDKECVYFC HLDIIWVNTP EHVVPYGLGS PRSKRALENL LPTKATDREN RCQCASQKDK KCWNFCQAGK ELRAEDIMEK DWNNHKKGKD CSKLGKKCIY QQLVRGRKIR RSSEEHLRQT RSETMRNSVK SSFHDPKLKG KPSRERYVTH NRAHW; Positive Samples: Mouse lung, Rat lung.
The protein encoded by this gene is proteolytically processed to release a secreted peptide termed endothelin 1. This peptide is a potent vasoconstrictor and is produced by vascular endothelial cells. Endothelin 1 also can affect the central nervous system. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
Endothelin 1 | |
Polyclonal | |
Unconjugated | |
EDN1 | |
ARCND3; Big endothelin 1; Big endothelin-1; Big ET; Big ET 1; Big ET1; BigET; EDN1; endothelin 1; endothelin-1; Et; ET1; ET-1; HDLCQ7; PPET1; ppET-1; preproendothelin; preproendothelin-1; preproET; QME | |
Rabbit | |
Affinity Chromatography | |
RUO | |
13614, 1906, 24323 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
P05305, P22387, P22388 | |
EDN1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 18-212 of human Endothelin 1 (NP_0019463). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.