Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Endothelin 1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585430
Description
Keep as concentrated solution. Positive Control: HUVEC, transfected 293T whole cell extracts. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is proteolytically processed to release a secreted peptide termed endothelin 1. This peptide is a potent vasoconstrictor and is produced by vascular endothelial cells. Endothelin 1 also can affect the central nervous system. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
Endothelin 1 | |
Polyclonal | |
Unconjugated | |
EDN1 | |
ARCND3; Big endothelin 1; Big endothelin-1; Big ET; Big ET 1; Big ET1; BigET; EDN1; endothelin 1; endothelin-1; Et; ET1; ET-1; HDLCQ7; PPET1; ppET-1; preproendothelin; preproendothelin-1; preproET; QME | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1906 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.2 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P05305 | |
EDN1 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Endothelin 1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction