Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Endothelin-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30353320UL
Description
Endothelin-1 Polyclonal specifically detects Endothelin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
Endothelin-1 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, KnockDown | |
Unconjugated | |
endothelin 1, endothelin-1, ET1, HDLCQ7, PPET1, Preproendothelin-1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 18-212 of human Endothelin-1 (NP_001946.3). | |
20 μL | |
Cell Cycle and Replication | |
1906 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction